missing translation for 'onlineSavingsMsg'
Learn More

2-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-4812725
85.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
51285-26-8
C6H7N3·ClH
157.6
GMHCEDDZKAYPLB-UHFFFAOYSA-N
picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
12403615
pyridine-2-carboximidamide;hydrochloride
C1=CC=NC(=C1)C(=N)N.Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
51285-26-8
C6H7N3·ClH
157.6
GMHCEDDZKAYPLB-UHFFFAOYSA-N
picolinimidamide hydrochloride, pyridine-2-carboximidamide hydrochloride, 2-amidinopyridine hydrochloride, pyridine-2-carboxamidine hydrochloride, picolinamidine hydrochloride, picolinamidine hcl, picolinimidamide hcl, pyridine-2-carboximidamide, hcl, 2-amidinopyridine, 2-amidinopyridinium chloride
12403615
pyridine-2-carboximidamide;hydrochloride
C1=CC=NC(=C1)C(=N)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 146.0°C to 148.0°C
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 2-Amidinopyridine Hydrochloride
compliance-icons
Product Identifier
  • 2-Amidinopyridine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'