missing translation for 'onlineSavingsMsg'
Learn More

2-Allyloxyethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-4812724
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
111-45-5
C5H10O2
102.13
MFCD00020607
GCYHRYNSUGLLMA-UHFFFAOYSA-N
2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether
8116
2-prop-2-enoxyethanol
OCCOCC=C
This item is not returnable. View return policy
111-45-5
C5H10O2
102.13
MFCD00020607
GCYHRYNSUGLLMA-UHFFFAOYSA-N
2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether
8116
2-prop-2-enoxyethanol
OCCOCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9550g/mL
Boiling Point 159.0°C
Flash Point 66°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHCH2OCH2CH2OH
Quantity 250 mL
Specific Gravity 0.955
Solubility Information Solubility in water: soluble
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material 2-Allyloxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Allyloxyethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'