missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-3-hydroxybenzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3880485
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
79950-42-8
C10H10O2
162.19
MFCD00100646
QVHRAGBOMUXWRI-UHFFFAOYSA-N
2-allyl-3-hydroxybenzaldehyde, 3-hydroxy-2-prop-2-en-1-yl benzaldehyde, 2-allyl-3hydroxybenzaldehyde, 2-allyl-3-hydroxy-benzaldehyde, 3-hydroxy-2-allyl benzaldehyde, 3-oxidanyl-2-prop-2-enyl-benzaldehyde, 3-hydroxy-2-prop-2-en-1-ylbenzaldehyde, benzaldehyde, 3-hydroxy-2-2-propenyl, benzaldehyde,3-hydroxy-2-2-propen-1-yl, 2-allyl-3-hydroxybenzaldehyde, containing 200ppm mehq
2775156
3-hydroxy-2-prop-2-enylbenzaldehyde
OC1=CC=CC(C=O)=C1CC=C
This item is not returnable. View return policy
79950-42-8
C10H10O2
162.19
MFCD00100646
QVHRAGBOMUXWRI-UHFFFAOYSA-N
2-allyl-3-hydroxybenzaldehyde, 3-hydroxy-2-prop-2-en-1-yl benzaldehyde, 2-allyl-3hydroxybenzaldehyde, 2-allyl-3-hydroxy-benzaldehyde, 3-hydroxy-2-allyl benzaldehyde, 3-oxidanyl-2-prop-2-enyl-benzaldehyde, 3-hydroxy-2-prop-2-en-1-ylbenzaldehyde, benzaldehyde, 3-hydroxy-2-2-propenyl, benzaldehyde,3-hydroxy-2-2-propen-1-yl, 2-allyl-3-hydroxybenzaldehyde, containing 200ppm mehq
2775156
3-hydroxy-2-prop-2-enylbenzaldehyde
OC1=CC=CC(C=O)=C1CC=C
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 2-Allyl-3-hydroxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'