missing translation for 'onlineSavingsMsg'
Learn More

2-Acrylamido-2-methylpropanesulfonic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10570934
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15214-89-8
C7H13NO4S
207.24
MFCD00007522
XHZPRMZZQOIPDS-UHFFFAOYSA-N
2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
65360
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
CC(C)(CS(O)(=O)=O)NC(=O)C=C
This item is not returnable. View return policy
15214-89-8
C7H13NO4S
207.24
MFCD00007522
XHZPRMZZQOIPDS-UHFFFAOYSA-N
2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
65360
2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
CC(C)(CS(O)(=O)=O)NC(=O)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 195.0°C
Color White
Density 160g/mL
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH2CHCONHC(CH3)2CH2SO3H
Quantity 250 g
Solubility Information Solubility in water: 1500g/L (20°C)
Formula Weight 207.25
Percent Purity 97%
Physical Form Powder, Chunks or Chips
Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Acrylamido-2-methylpropanesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'