missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylquinoxaline, 97%, Thermo Scientific Chemicals

Catalog Number p-7029237
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
25594-62-1
C10H8N2O
172.187
MFCD07808559
SWGILLQQUULMJB-UHFFFAOYSA-N
2-acetylquinoxaline, 1-quinoxalin-2-yl ethanone, ethanone, 1-2-quinoxalinyl, acmc-1cknl, 1-quinoxalin-2-yl ethan-1-one
11105814
1-quinoxalin-2-ylethanone
CC(=O)C1=NC2=CC=CC=C2N=C1
This item is not returnable. View return policy
25594-62-1
C10H8N2O
172.187
MFCD07808559
SWGILLQQUULMJB-UHFFFAOYSA-N
2-acetylquinoxaline, 1-quinoxalin-2-yl ethanone, ethanone, 1-2-quinoxalinyl, acmc-1cknl, 1-quinoxalin-2-yl ethan-1-one
11105814
1-quinoxalin-2-ylethanone
CC(=O)C1=NC2=CC=CC=C2N=C1

2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 76°C to 79°C
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 2-Acetylquinoxaline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'