missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylcyclohexanone, 97%, Thermo Scientific Chemicals

Catalog Number p-49484
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
874-23-7
C8H12O2
140.18
MFCD00001633
OEKATORRSPXJHE-UHFFFAOYNA-N
2-acetylcyclohexanone, cyclohexanone, 2-acetyl, 2-acetyl-1-cyclohexanone, .alpha.-acetylcyclohexanone, 2-acetyl cyclohexanone, 2-acetyl-cyclohexanone, cyclohexanone,2-acetyl, 1-acetyl-2-oxocyclohexane, alpha-acetylcyclohexanone, acetylcyclohexanone
13400
2-acetylcyclohexan-1-one
CC(=O)C1CCCCC1=O
This item is not returnable. View return policy
874-23-7
C8H12O2
140.18
MFCD00001633
OEKATORRSPXJHE-UHFFFAOYNA-N
2-acetylcyclohexanone, cyclohexanone, 2-acetyl, 2-acetyl-1-cyclohexanone, .alpha.-acetylcyclohexanone, 2-acetyl cyclohexanone, 2-acetyl-cyclohexanone, cyclohexanone,2-acetyl, 1-acetyl-2-oxocyclohexane, alpha-acetylcyclohexanone, acetylcyclohexanone
13400
2-acetylcyclohexan-1-one
CC(=O)C1CCCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0630g/mL
Boiling Point 224°C to 226°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H9(=O)
Refractive Index 1.5090 to 1.511
Quantity 5 g
Beilstein 07, 559
Specific Gravity 1.063
Formula Weight 140.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Acetylcyclohexanone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'