missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Acetylanthracene, 97%

Brand:  Alfa Aesar™ 044751.06

Code : NEW

Additional Details : CAS Number : 10210-32-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11479990

  • £132.08 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Acetylanthracene
C16H12O
MFCD00156727
10210-32-9
220.27

Specifications

2-Acetylanthracene
Powder
C16H12O
MFCD00156727
5g
10210-32-9
97%
220.27
188° to 192°C
Videos
SDS