missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific™

Catalog Number 15409025
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
81885-67-8
C11H14N2O
190.246
MFCD10016667
XWVFMJLNNGXNSG-UHFFFAOYSA-N
1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethanone, 2-acetyl-1,2,3,4-tetrahydroisoquinolin-7-amine, 2-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 7-isoquinolinamine, 2-acetyl-1,2,3,4-tetrahydro, 1-7-amino-3,4-dihydro-1h-isoquinolin-2-yl ethanone, n-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl ethanone, 1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethan-1-one, ethanone, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl, 1-7-amino-1,2,3,4-tetrahydroisoquinolin-2-yl ethan-1-one
12811352
1-(7-amino-3,4-dihydro-1H-isoquinolin-2-yl)ethanone
CC(=O)N1CCC2=C(C1)C=C(C=C2)N
This item is not returnable. View return policy
81885-67-8
C11H14N2O
190.246
MFCD10016667
XWVFMJLNNGXNSG-UHFFFAOYSA-N
1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethanone, 2-acetyl-1,2,3,4-tetrahydroisoquinolin-7-amine, 2-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 7-isoquinolinamine, 2-acetyl-1,2,3,4-tetrahydro, 1-7-amino-3,4-dihydro-1h-isoquinolin-2-yl ethanone, n-acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl ethanone, 1-7-amino-3,4-dihydroisoquinolin-2 1h-yl ethan-1-one, ethanone, 1-7-amino-3,4-dihydro-2 1h-isoquinolinyl, 1-7-amino-1,2,3,4-tetrahydroisoquinolin-2-yl ethan-1-one
12811352
1-(7-amino-3,4-dihydro-1H-isoquinolin-2-yl)ethanone
CC(=O)N1CCC2=C(C1)C=C(C=C2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 107°C to 111°C
Color Brown
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 190.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Acetyl-7-amino-1,2,3,4-tetrahydroisoquinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'