missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog Number p-49452
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3900-45-6
C13H12O2
200.24
MFCD00021643
GGWCZBGAIGGTDA-UHFFFAOYSA-N
2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
77506
1-(6-methoxynaphthalen-2-yl)ethanone
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC
This item is not returnable. View return policy
3900-45-6
C13H12O2
200.24
MFCD00021643
GGWCZBGAIGGTDA-UHFFFAOYSA-N
2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
77506
1-(6-methoxynaphthalen-2-yl)ethanone
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 108.0°C to 109.0°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC10H6COCH3
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Acetyl-6-methoxynaphthalene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetyl-6-methoxynaphthalene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'