missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylthiophene 98%, ACROS Organics™

Catalog Number p-3211570
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13679-74-8
C7H8OS
140.20
MFCD00014529
YOSDTJYMDAEEAZ-UHFFFAOYSA-N
2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
83655
1-(5-methylthiophen-2-yl)ethan-1-one
CC(=O)C1=CC=C(C)S1
This item is not returnable. View return policy
13679-74-8
C7H8OS
140.20
MFCD00014529
YOSDTJYMDAEEAZ-UHFFFAOYSA-N
2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
83655
1-(5-methylthiophen-2-yl)ethan-1-one
CC(=O)C1=CC=C(C)S1
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 140.2
Percent Purity 98%
Physical Form Low Melting Crystalline Mass, Powder or Crystals
Chemical Name or Material 2-Acetyl-5-methylthiophene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetyl-5-methylthiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'