missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-methylthiophene, 98%, Thermo Scientific Chemicals

Catalog Number p-7081321
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
13679-74-8
C7H8OS
140.20
MFCD00014529
YOSDTJYMDAEEAZ-UHFFFAOYSA-N
2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
83655
1-(5-methylthiophen-2-yl)ethanone
CC(=O)C1=CC=C(C)S1
This item is not returnable. View return policy
13679-74-8
C7H8OS
140.20
MFCD00014529
YOSDTJYMDAEEAZ-UHFFFAOYSA-N
2-acetyl-5-methylthiophene, 1-5-methylthiophen-2-yl ethanone, ketone, methyl 5-methyl-2-thienyl, 1-5-methyl-2-thienyl ethan-1-one, 2-methyl-5-acetylthiophene, ethanone, 1-5-methyl-2-thienyl, methylthienylcetone, 1-5-methyl-2-thienyl ethanone, 5-methyl-2-acetylthiophene, methyl 5-methyl-2-thienyl ketone
83655
1-(5-methylthiophen-2-yl)ethanone
CC(=O)C1=CC=C(C)S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 24°C to 28°C
Density 1.119
Boiling Point 65°C to 67°C (1 mmHg)
Refractive Index 1.5625
Quantity 5 g
Beilstein 110854
Sensitivity Light sensitive
Formula Weight 140.2
Percent Purity 98%
Chemical Name or Material 2-Acetyl-5-methylthiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'