missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Acetyl-5-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62931.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468016

  • £352.70 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Acetyl-5-fluorobenzeneboronic acid pinacol ester
264.1
2-Acetyl-5-fluorophenylboronic acid pinacol ester

Specifications

2-Acetyl-5-fluorobenzeneboronic acid pinacol ester
Solid
264.1
1g
1809548-73-9
96%
2-Acetyl-5-fluorophenylboronic acid pinacol ester
Videos
SDS