missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-bromothiophene, 99%, Thermo Scientific Chemicals

Catalog Number 10567763
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
5370-25-2
C6H5BrOS
205.07
MFCD00014528
IGBZCOWXSCWSHO-UHFFFAOYSA-N
2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
79335
1-(5-bromothiophen-2-yl)ethanone
CC(=O)C1=CC=C(S1)Br
This item is not returnable. View return policy
5370-25-2
C6H5BrOS
205.07
MFCD00014528
IGBZCOWXSCWSHO-UHFFFAOYSA-N
2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
79335
1-(5-bromothiophen-2-yl)ethanone
CC(=O)C1=CC=C(S1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Yellow
Boiling Point 103°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 205.07
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 2-Acetyl-5-bromothiophene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'