missing translation for 'onlineSavingsMsg'
Learn More

2'-Acetonaphthone, 99%, Thermo Scientific Chemicals

Catalog Number p-48339
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
93-08-3
C12H10O
170.21
MFCD00004108
XSAYZAUNJMRRIR-UHFFFAOYSA-N
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
7122
CHEBI:52364
1-naphthalen-2-ylethanone
CC(=O)C1=CC2=CC=CC=C2C=C1
This item is not returnable. View return policy
93-08-3
C12H10O
170.21
MFCD00004108
XSAYZAUNJMRRIR-UHFFFAOYSA-N
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
7122
CHEBI:52364
1-naphthalen-2-ylethanone
CC(=O)C1=CC2=CC=CC=C2C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Color White
Boiling Point 300.0°C to 301.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C10H7COCH3
Quantity 100 g
Beilstein 07, 402
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 99%
Physical Form Fine Crystalline Powder and Chunks
Chemical Name or Material 2'-Acetonaphthone, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2'-Acetonaphthone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'