2-Acetamidofluorene, 95%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 102300050

5GR 2-Acetamidofluorene, 95%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 53-96-3 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 10528100

Quantity Price
1 £ 42.4 / 5g
EU Stock 2
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Acetamidofluorene
CAS 53-96-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15H13NO
Formula Weight 223.27
MDL Number MFCD00001116
Synonym N-(2-Fluorenyl)acetamide

Specification

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 223.27
Quantity 5g
Color White to yellow-brown
Beilstein 12, 1331
Merck Index 15, 4189
CAS Min % 94.0
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 53-96-3
Assay Percent Range 95%
MDL Number MFCD00001116
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause cancer.
Harmful if swallowed.
Molecular Formula C15H13NO
CAS Max % 100.0
Chemical Name or Material 2-Acetamidofluorene
Synonym N-(2-Fluorenyl)acetamide
Melting Point 192° to 196°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
May cause cancer.
Harmful if swallowed.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.