missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-5-bromopyridine, 98%, Thermo Scientific™

Catalog Number 11916441
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7169-97-3
C7H7BrN2O
215.05
MJFCOXATGBYERZ-UHFFFAOYSA-N
2-acetamido-5-bromopyridine, n-5-bromopyridin-2-yl acetamide, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridyl acetamide, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-acetylamino-5-bromopyridine, pubchem5546
293097
N-(5-bromopyridin-2-yl)acetamide
CC(=O)NC1=NC=C(C=C1)Br
This item is not returnable. View return policy
7169-97-3
C7H7BrN2O
215.05
MJFCOXATGBYERZ-UHFFFAOYSA-N
2-acetamido-5-bromopyridine, n-5-bromopyridin-2-yl acetamide, 2-acetylamino-5-bromopyridine, acetamide, n-5-bromo-2-pyridinyl, n-5-bromo-2-pyridyl acetamide, n-5-bromo-2-pyridinyl acetamide, n-5-bromo-pyridin-2-yl-acetamide, n-acetylamino-5-bromopyridine, pubchem5546
293097
N-(5-bromopyridin-2-yl)acetamide
CC(=O)NC1=NC=C(C=C1)Br
Melting Point 176°C to 178°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Acetamido-5-bromopyridine
compliance-icons
Product Identifier
  • 2-Acetamido-5-bromopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'