missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-47631
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
38985-79-4
C9H7BrNO3
257.06
MFCD00040904
QVABAFHRLMDDLM-UHFFFAOYSA-M
520935
CC(=O)NC1=CC=C(Br)C=C1C([O-])=O
This item is not returnable. View return policy
38985-79-4
C9H7BrNO3
257.06
MFCD00040904
QVABAFHRLMDDLM-UHFFFAOYSA-M
520935
CC(=O)NC1=CC=C(Br)C=C1C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 218°C to 223°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C(o)NHC6H3(Br)COOH
Quantity 5 g
Synonym 5-bromo-2-acetamidobenzoic acid, 2-acetamino-5-bromobenzoic acid, n-acetyl-5-bromo-anthranilic acid, 2-acetylamino-5-bromobenzoic acid, n-acetyl-5-bromoanthranilic acid, 2-acetamido-5-bromobenzoic acid, benzoic acid,2-acetylamino-5-bromo, benzoic acid, 2-acetylamino-5-bromo
IUPAC Name 2-acetamido-5-bromobenzoic acid
Formula Weight 258.07
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Acetamido-5-bromobenzoic acid, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'