missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, ACROS Organics™

Brand:  Acros Organics 102560250

Code : 29

Additional Details : CAS Number : 10036-64-3 Weight : 0.07500kg

Quantity 25g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10386160

  • £28.60 / 25g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

2-Acetamido-2-deoxy-α-D-glucopyranose
10036-64-3
100.0
99+%
221.21
N-Acetyl-α-D-glucosamine
99.5+%
99.0
≥99.0%
C8H15NO6
MFCD00064359

Specifications

2-Acetamido-2-deoxy-α-D-glucopyranose
10036-64-3
100.0
Powder
99+%
C8H15NO6
MFCD00064359
31, 170
N-Acetyl-α-D-glucosamine
0.5% max.
White to off-white
99.5+%
99.0
Authentic
≥99.0%
1.0% max. (105°C, 4 hrs)
221.21
Plastic bottle
10, 4318
+39° to +43° (20°C, 589nm) (c=2, water)
+ 41.00 (20.00°C c=2,H2O)
25g
Videos
SDS