missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,9-Dimethyl-1,10-phenanthroline hemihydrate, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Description
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
Solubility
It is soluble in methanol: 0.1 g/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.
2,9-Dimethyl-1,10-phenanthroline hemihydrate, is used as a reagent for the spectrophotometric determination of copper. It is used as an intermediate in medicine and chemical research.
Solubility
It is soluble in methanol: 0.1 g/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents.
Specifications
Melting Point | 160°C to 164°C |
Quantity | 5 g |
Beilstein | 153179 |
Merck Index | 14,6449 |
Solubility Information | It is soluble in methanol: 0.1g/mL. |
Formula Weight | 217.27 (208.26 Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | 2,9-Dimethyl-1,10-phenanthroline hemihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'