missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Di-tert-butylfluorene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used as an organic light emitting diode (OLED) intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an organic light emitting diode (OLED) intermediate.
Solubility
Soluble in toluene, hot methanol.
Notes
Store at -20°C. Avoid heat and oxidizing agents.
It is used as an organic light emitting diode (OLED) intermediate.
Solubility
Soluble in toluene, hot methanol.
Notes
Store at -20°C. Avoid heat and oxidizing agents.
Specifications
Melting Point | 121°C to 124°C |
Quantity | 1 g |
Solubility Information | Soluble in toluene,hot methanol. |
Formula Weight | 278.43 |
Percent Purity | 98% |
Chemical Name or Material | 2,7-Di-tert-butylfluorene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'