missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-4228890
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5431-44-7
C7H5NO2
135.12
PMWXGSWIOOVHEQ-UHFFFAOYSA-N
2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde #
79485
pyridine-2,6-dicarbaldehyde
C1=CC(=NC(=C1)C=O)C=O
This item is not returnable. View return policy
5431-44-7
C7H5NO2
135.12
PMWXGSWIOOVHEQ-UHFFFAOYSA-N
2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde #
79485
pyridine-2,6-dicarbaldehyde
C1=CC(=NC(=C1)C=O)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 123.3°C to 124.5°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 135.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 6-Pyridinedicarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Pyridinedicarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'