missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(6-Methyl-2-pyridyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62907.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437986

  • £99.64 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(6-Methyl-2-pyridyl)thiazolidine-4-carboxylic acid
224.28
MFCD26960683

Specifications

2-(6-Methyl-2-pyridyl)thiazolidine-4-carboxylic acid
Solid
224.28
1g
97%
MFCD26960683
Videos
SDS