missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpyridin-3-ol, 97%, Maybridge

£62.00 - £363.00

Chemical Identifiers

CAS 1122-43-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00228692
InChI Key AMQJAKKATFMFTR-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3-hydroxypyridine, 2,6-dimethyl-3-pyridinol, 2,6-dimethyl-3-oxypyridine, 3-pyridinol, 2,6-dimethyl, 3-hydroxy-2,6-lutidine, 2,6-dimethyl-pyridin-3-ol, 3-hydroxy-2,6-dimethylpyridine, pubchem22495, 2,6-dimethyl-3-pyridino, 2,6-dimethylpyridine-3-ol
PubChem CID 70732
IUPAC Name 2,6-dimethylpyridin-3-ol
SMILES CC1=NC(=C(C=C1)O)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10190932 SDS View Documents Maybridge
RH01494DA
1g £62.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 01-06-2020
Please to purchase this item. Need a web account? Register with us today!
10361615 SDS View Documents Maybridge
RH01494DE
5g £198.00
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-07-2020
Please to purchase this item. Need a web account? Register with us today!
10019341 SDS View Documents Maybridge
RH01494EA
10g £363.00
10g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1122-43-6
123.155
AMQJAKKATFMFTR-UHFFFAOYSA-N
70732
CC1=NC(=C(C=C1)O)C
C7H9NO
MFCD00228692
2,6-dimethyl-3-hydroxypyridine, 2,6-dimethyl-3-pyridinol, 2,6-dimethyl-3-oxypyridine, 3-pyridinol, 2,6-dimethyl, 3-hydroxy-2,6-lutidine, 2,6-dimethyl-pyridin-3-ol, 3-hydroxy-2,6-dimethylpyridine, pubchem22495, 2,6-dimethyl-3-pyridino, 2,6-dimethylpyridine-3-ol
2,6-dimethylpyridin-3-ol

Specifications

2, 6-Dimethylpyridin-3-ol
1g
97.0
C7H9NO
2,6-dimethyl-3-hydroxypyridine, 2,6-dimethyl-3-pyridinol, 2,6-dimethyl-3-oxypyridine, 3-pyridinol, 2,6-dimethyl, 3-hydroxy-2,6-lutidine, 2,6-dimethyl-pyridin-3-ol, 3-hydroxy-2,6-dimethylpyridine, pubchem22495, 2,6-dimethyl-3-pyridino, 2,6-dimethylpyridine-3-ol
CC1=NC(=C(C=C1)O)C
123.155
123.15
Amber glass bottle
1122-43-6
97%
MFCD00228692
AMQJAKKATFMFTR-UHFFFAOYSA-N
2,6-dimethylpyridin-3-ol
70732
97%