missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylpyridin-3-ol, 97%, Thermo Scientific™

Catalog Number 10361615
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
1122-43-6
C7H9NO
123.16
MFCD00228692
AMQJAKKATFMFTR-UHFFFAOYSA-N
2,6-dimethyl-3-hydroxypyridine, 2,6-dimethyl-3-pyridinol, 2,6-dimethyl-3-oxypyridine, 3-pyridinol, 2,6-dimethyl, 3-hydroxy-2,6-lutidine, 2,6-dimethyl-pyridin-3-ol, 3-hydroxy-2,6-dimethylpyridine, pubchem22495, 2,6-dimethyl-3-pyridino, 2,6-dimethylpyridine-3-ol
70732
2,6-dimethylpyridin-3-ol
CC1=CC=C(O)C(C)=N1
This item is not returnable. View return policy
1122-43-6
C7H9NO
123.16
MFCD00228692
AMQJAKKATFMFTR-UHFFFAOYSA-N
2,6-dimethyl-3-hydroxypyridine, 2,6-dimethyl-3-pyridinol, 2,6-dimethyl-3-oxypyridine, 3-pyridinol, 2,6-dimethyl, 3-hydroxy-2,6-lutidine, 2,6-dimethyl-pyridin-3-ol, 3-hydroxy-2,6-dimethylpyridine, pubchem22495, 2,6-dimethyl-3-pyridino, 2,6-dimethylpyridine-3-ol
70732
2,6-dimethylpyridin-3-ol
CC1=CC=C(O)C(C)=N1
CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethylpyridin-3-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'