missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylpiperazine, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | 108.0°C to 113.0°C |
Boiling Point | 162.0°C |
Flash Point | 44°C |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 25 g |
Beilstein | 23,I,8 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents |
Formula Weight | 114.19 |
Percent Purity | 98% |
Chemical Name or Material | 2, 6-Dimethylpiperazine |
Safety and Handling
Product Identifier
- 2,6-Dimethylpiperazine
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'