missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog Number p-7053754
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
62285-58-9
C9H12O
136.194
MFCD00128004
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl
186748
(2,6-dimethylphenyl)methanol
CC1=C(C(=CC=C1)C)CO
This item is not returnable. View return policy
62285-58-9
C9H12O
136.194
MFCD00128004
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl
186748
(2,6-dimethylphenyl)methanol
CC1=C(C(=CC=C1)C)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 136.19
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,6-Dimethylbenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'