missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number 10563531
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
632-46-2
C9H10O2
150.18
MFCD00002483
HCBHQDKBSKYGCK-UHFFFAOYSA-N
benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid
12439
CHEBI:64827
2,6-dimethylbenzoic acid
CC1=C(C(=CC=C1)C)C(=O)O
This item is not returnable. View return policy
632-46-2
C9H10O2
150.18
MFCD00002483
HCBHQDKBSKYGCK-UHFFFAOYSA-N
benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid
12439
CHEBI:64827
2,6-dimethylbenzoic acid
CC1=C(C(=CC=C1)C)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 114.0°C to 117.0°C
CAS Min % 98.5
Color Cream to White
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3COOH
Beilstein 01, 259
Formula Weight 150.18
Percent Purity 99%
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dimethylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'