missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog Number p-7021960
Click to view available options
Quantity:
100 g
500 g
1000 g
This item is not returnable. View return policy
87-62-7
C8H11N
121.18
MFCD00007747
UFFBMTHBGFGIHF-UHFFFAOYSA-N
2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
6896
CHEBI:28738
2,6-dimethylaniline
CC1=CC=CC(C)=C1N
This item is not returnable. View return policy
87-62-7
C8H11N
121.18
MFCD00007747
UFFBMTHBGFGIHF-UHFFFAOYSA-N
2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
6896
CHEBI:28738
2,6-dimethylaniline
CC1=CC=CC(C)=C1N

2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.

Solubility
Slightly miscible with water. Miscible with ethanol and diethyl ether.

Notes
It is sensitive to air and light. Incompatible withacids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates and halogens.
TRUSTED_SUSTAINABILITY
Melting Point 11°C
Density 0.981
Boiling Point 216°C
Quantity 100 g
Odor Characteristic
Refractive Index 1.56
UN Number UN1711
Beilstein 636332
Merck Index 14,10084
Solubility Information Slightly miscible with water. Miscible with ethanol and diethyl ether.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'