missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, Thermo Scientific™

Catalog Number p-2689861
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
269410-25-5
C14H21BO3
248.13
MFCD02093724
TYCKOBOJYNRIBO-UHFFFAOYSA-N
2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxy-3,5-dimethylphenylboronic acid pinacol ester, 3,5-dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-dimethyl-4-hydroxybenzeneboronic acid, pinacol ester, 4-hydroxy-3,5-dimethylphenylboronic acid, pinacol ester, 2,6-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2,6-dimethyl-4-4,4-5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-4-hydroxy-3,5-dimethylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2734649
2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CC1=CC(=CC(C)=C1O)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
269410-25-5
C14H21BO3
248.13
MFCD02093724
TYCKOBOJYNRIBO-UHFFFAOYSA-N
2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxy-3,5-dimethylphenylboronic acid pinacol ester, 3,5-dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-dimethyl-4-hydroxybenzeneboronic acid, pinacol ester, 4-hydroxy-3,5-dimethylphenylboronic acid, pinacol ester, 2,6-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2,6-dimethyl-4-4,4-5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-4-hydroxy-3,5-dimethylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2734649
2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CC1=CC(=CC(C)=C1O)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 104.0°C to 109.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 248.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'