missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dimethyl-3,5-heptanedione, 98+%

Metal chelating agent; preparation of metal diketonates for MOCVD

Brand:  Alfa Aesar™ 042005.03

Code : NEW

Additional Details : CAS Number : 18362-64-6 Weight : 0.00100kg Transport : UN number : 1224 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11385017

  • £42.30 / 1g


Estimated Shipment date
from Supplier 13-02-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Dimethyl-3,5-heptanedione
156.22
UN1224
18362-64-6
MFCD00015040

Specifications

2,6-Dimethyl-3,5-heptanedione
Liquid
≥98%
MFCD00015040
1g
18362-64-6
0.91g/mL
156.22
UN1224
Videos
SDS