missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxytoluene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
Description
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 37°C to 41°C |
Boiling Point | 222°C |
Flash Point | 96°C (204°F) |
Odor | Aromatic |
Molecular Formula | C4H7ClMg |
MDL Number | MFCD00674003 |
Quantity | 25 g |
Beilstein | 2045327 |
Synonym | methallylmagnesium chloride, 2-methylallylmagnesium chloride, magnesium, chloro 2-methyl-2-propenyl, methallyl magnesium chloride, methallyl-magnesium chloride, 2-methylallyl magnesium chloride, bjvfgwybolmuem-uhfffaoysa-m, 2-methylallylmagnesium chloride solution, 2-methyl-2-propenyl magnesium chloride, 2-methyl-2-propen-1-yl magnesium chloride |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'