missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7029779
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
214360-59-5
C13H20BNO4
265.12
MFCD03788240
VZGDCNFVMWRPOW-UHFFFAOYSA-N
2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxypyridine-3-boronic acid pinacol ester, 2,6-dimethoxypyridine-5-boronic acid pinacol ester, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2,6-dimethoxypyridine-3-boronic acid, pinacol ester, 2,6-dimethoxy-5 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2,6-dimethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxabo
2758487
2,6-dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1
This item is not returnable. View return policy
214360-59-5
C13H20BNO4
265.12
MFCD03788240
VZGDCNFVMWRPOW-UHFFFAOYSA-N
2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxypyridine-3-boronic acid pinacol ester, 2,6-dimethoxypyridine-5-boronic acid pinacol ester, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2,6-dimethoxypyridine-3-boronic acid, pinacol ester, 2,6-dimethoxy-5 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2,6-dimethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxabo
2758487
2,6-dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 39°C to 42°C
Quantity 250 mg
Formula Weight 265.12
Percent Purity 95%
Chemical Name or Material 2,6-Dimethoxypyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'