missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxybenzonitrile, 97%, Thermo Scientific™

Catalog Number p-3757738
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16932-49-3
C9H9NO2
163.18
MFCD00001788
XHAHKSSLDJIEDH-UHFFFAOYSA-N
benzonitrile, 2,6-dimethoxy, 2,6-dimethoxy-benzonitrile, 2,6-dimethoxybenzenecarbonitrile, pubchem4772, 2.6-dimethoxybenzonitrile, benzonitrile,2,6-dimethoxy, 2-10-00-00260 beilstein handbook reference, 2,6-dimethoxybenzonitrile
85648
2,6-dimethoxybenzonitrile
COC1=C(C(=CC=C1)OC)C#N
This item is not returnable. View return policy
16932-49-3
C9H9NO2
163.18
MFCD00001788
XHAHKSSLDJIEDH-UHFFFAOYSA-N
benzonitrile, 2,6-dimethoxy, 2,6-dimethoxy-benzonitrile, 2,6-dimethoxybenzenecarbonitrile, pubchem4772, 2.6-dimethoxybenzonitrile, benzonitrile,2,6-dimethoxy, 2-10-00-00260 beilstein handbook reference, 2,6-dimethoxybenzonitrile
85648
2,6-dimethoxybenzonitrile
COC1=C(C(=CC=C1)OC)C#N
CAS Min % 96.0
CAS Max % 100.0
Melting Point 119°C to 123°C
Color Beige
Boiling Point 310°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CN
Quantity 10 g
Beilstein 10, 388
Formula Weight 163.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dimethoxybenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethoxybenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'