missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-dihydroxyisonicotinic acid, Thermo Scientific™
Click to view available options
Quantity:
50 g
100 g
Specifications
Assay Percent Range | 90% |
Linear Formula | (HO)2(C5H2N)COOH |
Quantity | 50 g |
Formula Weight | 155.11 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 2, 6-Dihydroxyisonicotinic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'