missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dihydroxy-4-methylpyridine, 98+%

Catalog Number p-7030296
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
4664-16-8
C6H7NO2
125.127
MFCD00040464
JJHVYGVVMBYCMQ-UHFFFAOYSA-N
6-hydroxy-4-methylpyridin-2 1h-one, 6-hydroxy-4-methyl-2-pyridone, 2,6-dihydroxy-4-methylpyridine, unii-0g0nnm04dt, 4-methylpyridine-2,6-diol, 0g0nnm04dt, 6-hydroxy-4-methyl-1,2-dihydropyridin-2-one, 2 1h-pyridinone, 6-hydroxy-4-methyl, acmc-209uzm, 4-methyl-2,6-dihydroxypyridine
78395
6-hydroxy-4-methyl-1H-pyridin-2-one
CC1=CC(=O)NC(=C1)O
This item is not returnable. View return policy
4664-16-8
C6H7NO2
125.127
MFCD00040464
JJHVYGVVMBYCMQ-UHFFFAOYSA-N
6-hydroxy-4-methylpyridin-2 1h-one, 6-hydroxy-4-methyl-2-pyridone, 2,6-dihydroxy-4-methylpyridine, unii-0g0nnm04dt, 4-methylpyridine-2,6-diol, 0g0nnm04dt, 6-hydroxy-4-methyl-1,2-dihydropyridin-2-one, 2 1h-pyridinone, 6-hydroxy-4-methyl, acmc-209uzm, 4-methyl-2,6-dihydroxypyridine
78395
6-hydroxy-4-methyl-1H-pyridin-2-one
CC1=CC(=O)NC(=C1)O
Applications
In order to synthesize macrocycles containing pyridine, 2,6-his-(bromomethyl)pyridine was prepared by bromonating 2,6-di (hydroxymethyl)pyridine.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 195°C to 198°C
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 125.13
Percent Purity ≥98%
Chemical Name or Material 2,6-Dihydroxy-4-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'