missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific™

Catalog Number 15491619
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
155601-65-3
C6H3F2NO
143.093
MFCD08277633
ZYCCUPSLEFLBNR-UHFFFAOYSA-N
2,6-difluoronicotinaldehyde, 2,6-difluoropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro, 2,6-difluoro-3-nicotinaldehyde, pubchem18894, 2,6-difluoro-3-formylpyridine, 2,6-difluoro-pyridine-3-carbaldehyde, 2,6-difluoro-3-pyridinecarboxaldehyde, 2,6-bis fluoranyl pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro-9ci
10701929
2,6-difluoropyridine-3-carbaldehyde
C1=CC(=NC(=C1C=O)F)F
This item is not returnable. View return policy
155601-65-3
C6H3F2NO
143.093
MFCD08277633
ZYCCUPSLEFLBNR-UHFFFAOYSA-N
2,6-difluoronicotinaldehyde, 2,6-difluoropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro, 2,6-difluoro-3-nicotinaldehyde, pubchem18894, 2,6-difluoro-3-formylpyridine, 2,6-difluoro-pyridine-3-carbaldehyde, 2,6-difluoro-3-pyridinecarboxaldehyde, 2,6-bis fluoranyl pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro-9ci
10701929
2,6-difluoropyridine-3-carbaldehyde
C1=CC(=NC(=C1C=O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 19°C to 20°C
Density 1.378 g/mL
Boiling Point 64°C to 66°C (15 mmHg)
Refractive Index 1.492
Quantity 1 g
Beilstein 7757317
Sensitivity Air sensitive
Formula Weight 143.09
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,6-Difluoropyridine-3-carboxaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'