missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Difluoropyridine-3-carboxaldehyde, 95%

Brand:  Alfa Aesar™ H64422.06

Code : NEW

Additional Details : CAS Number : 155601-65-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401629

  • £96.24 / 5g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

155601-65-3
143.093
ZYCCUPSLEFLBNR-UHFFFAOYSA-N
10701929
C1=CC(=NC(=C1C=O)F)F
C6H3F2NO
MFCD08277633
2,6-difluoronicotinaldehyde, 2,6-difluoropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro, 2,6-difluoro-3-nicotinaldehyde, pubchem18894, 2,6-difluoro-3-formylpyridine, 2,6-difluoro-pyridine-3-carbaldehyde, 2,6-difluoro-3-pyridinecarboxaldehyde, 2,6-bis fluoranyl pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro-9ci
2,6-difluoropyridine-3-carbaldehyde

Specifications

2,6-Difluoropyridine-3-carboxaldehyde
1.492
64°C to 66°C (15mmHg)
5g
C6H3F2NO
7757317
ZYCCUPSLEFLBNR-UHFFFAOYSA-N
2,6-difluoropyridine-3-carbaldehyde
10701929
Liquid
1.378 g/mL
Air sensitive
19°C to 20°C
155601-65-3
MFCD08277633
2,6-difluoronicotinaldehyde, 2,6-difluoropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro, 2,6-difluoro-3-nicotinaldehyde, pubchem18894, 2,6-difluoro-3-formylpyridine, 2,6-difluoro-pyridine-3-carbaldehyde, 2,6-difluoro-3-pyridinecarboxaldehyde, 2,6-bis fluoranyl pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 2,6-difluoro-9ci
C1=CC(=NC(=C1C=O)F)F
143.093
143.09
95%
Videos
SDS Documents
Certificates