missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorophenol, 98+%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,6-Difluorophenol undergoes oxidative polymerization in the presence of the Fe-N,N′-bis(salicylidene)ethylenediamine (salen) complex (catalyst) and hydrogen peroxide (oxidizing agent) to give poly(2,6-difluoro-1,4-phenylene oxide). It is also used as pharmaceutical intermediates.
Solubility
Slightly soluble in water. Soluble in ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2,6-Difluorophenol undergoes oxidative polymerization in the presence of the Fe-N,N′-bis(salicylidene)ethylenediamine (salen) complex (catalyst) and hydrogen peroxide (oxidizing agent) to give poly(2,6-difluoro-1,4-phenylene oxide). It is also used as pharmaceutical intermediates.
Solubility
Slightly soluble in water. Soluble in ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 36°C to 40°C |
Density | 1.27 |
Boiling Point | 59°C to 61°C (17 mmHg) |
Flash Point | 58°C (136°F) |
Quantity | 5 g |
UN Number | UN2921 |
Beilstein | 2043613 |
Solubility Information | Slightly soluble in water. Soluble in ethanol. |
Formula Weight | 130.09 |
Percent Purity | ≥98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'