missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzylamine, 97+%, Thermo Scientific™

Catalog Number p-7023525
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
69385-30-4
C7H8F2N
144.14
MFCD00010144
PQCUDKMMPTXMAL-UHFFFAOYSA-O
123563
[NH3+]CC1=C(F)C=CC=C1F
This item is not returnable. View return policy
69385-30-4
C7H8F2N
144.14
MFCD00010144
PQCUDKMMPTXMAL-UHFFFAOYSA-O
123563
[NH3+]CC1=C(F)C=CC=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.197
Boiling Point 174°C to 177°C
Flash Point 65°C (149°F)
Refractive Index 1.493
Quantity 1 g
UN Number UN2735
Beilstein 2716771
Sensitivity Air Sensitive
Synonym 2,6-difluorobenzylamine, 2,6-difluorophenyl methanamine, 2,6-difluorobenzyl amine, benzenemethanamine, 2,6-difluoro, 1-2,6-difluorophenyl methanamine, 2,6-difluorophenyl methylamine, 2, 6-difluorobenzylamine, pubchem2235, 2,6-difluoro-benzylamine, acmc-1b2iy
IUPAC Name (2,6-difluorophenyl)methanamine
Formula Weight 143.14
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluorobenzylamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'