missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-3757667
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
437-81-0
C7H4F2O
142.11
MFCD00010293
SOWRUJSGHKNOKN-UHFFFAOYSA-N
2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
136284
2,6-difluorobenzaldehyde
C1=CC(=C(C(=C1)F)C=O)F
This item is not returnable. View return policy
437-81-0
C7H4F2O
142.11
MFCD00010293
SOWRUJSGHKNOKN-UHFFFAOYSA-N
2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
136284
2,6-difluorobenzaldehyde
C1=CC(=C(C(=C1)F)C=O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 15°C to 17°C
Color Yellow
Density 1.3100g/mL
Boiling Point 82°C to 84°C (15.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3COH
Refractive Index 1.5060 to 1.509
Quantity 5 g
Specific Gravity 1.31
Formula Weight 142.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'