missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Difluoroacetophenone, 98%, Thermo Scientific™

Catalog Number 10716305
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13670-99-0
C8H6F2O
156.13
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
83643
1-(2,6-difluorophenyl)ethanone
CC(=O)C1=C(F)C=CC=C1F
This item is not returnable. View return policy
13670-99-0
C8H6F2O
156.13
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
83643
1-(2,6-difluorophenyl)ethanone
CC(=O)C1=C(F)C=CC=C1F
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 76.0°C to 79.0°C (15.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3COCH3
Refractive Index 1.48
Quantity 1 g
Specific Gravity 1.19
Formula Weight 156.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2', 6'-Difluoroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'