missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Difluoroacetophenone, 98%, ACROS Organics™

Brand:  Acros Organics 214290050

Code : 29

Additional Details : CAS Number : 13670-99-0 Weight : 0.06000kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10071694

  • £65.98 / 5g

Estimated Shipment
15-12-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

13670-99-0
156.13
VGIIILXIQLXVLC-UHFFFAOYSA-N
83643
CC(=O)C1=C(C=CC=C1F)F
C8H6F2O
MFCD00000328
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
1-(2,6-difluorophenyl)ethanone

Specifications

2', 6'-Difluoroacetophenone
76°C
Glass bottle
1.19
Colorless to Yellow
13670-99-0
100.0
C8H6F2O
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
1-(2,6-difluorophenyl)ethanone
83643
Liquid
1.1900g/mL
Authentic
1.48
76.0°C to 79.0°C (15.0mmHg)
5g
97.5
97.5% min. (GC)
F2C6H3COCH3
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
CC(=O)C1=C(C=CC=C1F)F
156.13
156.13
98%
Videos
SDS Documents