missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Difluoro-N-(3-fluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59831.03

Code : NEW

Additional Details : CAS Number : 937635-68-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406258

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Difluoro-N-(3-fluorophenyl)benzamide
251.2
23653386
937635-68-2
MFCD07820289

Specifications

2,6-Difluoro-N-(3-fluorophenyl)benzamide
1g
Solid
251.2
23653386
White
937635-68-2
97%
MFCD07820289
Videos
SDS