missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Difluoro-N-(2-fluoro-5-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59598.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15425126

  • £358.49 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Difluoro-N-(2-fluoro-5-methylphenyl)benzamide
265.23
MFCD09072119

Specifications

2,6-Difluoro-N-(2-fluoro-5-methylphenyl)benzamide
Solid
265.23
1g
97%
MFCD09072119
Videos
SDS