missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2,6-Difluoro-N-(2,5-difluorophenyl)benzamide, 97%
Brand: Alfa Aesar™ H59620.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2,6-Difluoro-N-(2,5-difluorophenyl)benzamide | |
269.19 | |
MFCD07830378 |
Specifications
2,6-Difluoro-N-(2,5-difluorophenyl)benzamide | |
Solid | |
269.19 |
1g | |
97% | |
MFCD07830378 |