missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Difluoro-N-(2,4-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59196.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15425438

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Difluoro-N-(2,4-difluorophenyl)benzamide
269.19
MFCD00115791

Specifications

2,6-Difluoro-N-(2,4-difluorophenyl)benzamide
Solid
269.19
1g
97%
MFCD00115791
Videos
SDS