missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-methoxybenzyl bromide, 97%, Thermo Scientific™

Catalog Number 11426030
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
94278-68-9
C8H7BrF2O
237.044
MFCD04115925
JCRQCVDDBZVTMB-UHFFFAOYSA-N
2,6-difluoro-4-methoxybenzyl bromide, 2-bromomethyl-1,3-difluoro-5-methoxybenzene, 2-bromomethyl-1,3-difluoro-5-methoxy-benzene, 2-bromomethyl-1,3-bis fluoranyl-5-methoxy-benzene
3354272
2-(bromomethyl)-1,3-difluoro-5-methoxybenzene
COC1=CC(=C(C(=C1)F)CBr)F
This item is not returnable. View return policy
94278-68-9
C8H7BrF2O
237.044
MFCD04115925
JCRQCVDDBZVTMB-UHFFFAOYSA-N
2,6-difluoro-4-methoxybenzyl bromide, 2-bromomethyl-1,3-difluoro-5-methoxybenzene, 2-bromomethyl-1,3-difluoro-5-methoxy-benzene, 2-bromomethyl-1,3-bis fluoranyl-5-methoxy-benzene
3354272
2-(bromomethyl)-1,3-difluoro-5-methoxybenzene
COC1=CC(=C(C(=C1)F)CBr)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 43°C to 46°C
Quantity 5 g
UN Number UN3261
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-4-methoxybenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'