missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Difluoro-4-methoxybenzyl bromide, 97%

Brand:  Alfa Aesar™ H26576.03

Code : NEW

Additional Details : CAS Number : 94278-68-9 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11416030

  • £58.00 / 1g

Estimated Shipment date
from Supplier 04-03-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

94278-68-9
237.044
JCRQCVDDBZVTMB-UHFFFAOYSA-N
3354272
COC1=CC(=C(C(=C1)F)CBr)F
C8H7BrF2O
MFCD04115925
2,6-difluoro-4-methoxybenzyl bromide, 2-bromomethyl-1,3-difluoro-5-methoxybenzene, 2-bromomethyl-1,3-difluoro-5-methoxy-benzene, 2-bromomethyl-1,3-bis fluoranyl-5-methoxy-benzene
2-(bromomethyl)-1,3-difluoro-5-methoxybenzene

Specifications

2,6-Difluoro-4-methoxybenzyl bromide
1g
C8H7BrF2O
UN3261
JCRQCVDDBZVTMB-UHFFFAOYSA-N
2-(bromomethyl)-1,3-difluoro-5-methoxybenzene
3354272
97%
43°C to 46°C
94278-68-9
MFCD04115925
2,6-difluoro-4-methoxybenzyl bromide, 2-bromomethyl-1,3-difluoro-5-methoxybenzene, 2-bromomethyl-1,3-difluoro-5-methoxy-benzene, 2-bromomethyl-1,3-bis fluoranyl-5-methoxy-benzene
COC1=CC(=C(C(=C1)F)CBr)F
237.044
237.04
Videos
SDS Documents