missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081814
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
50709-36-9
C6H7Cl3N2
213.486
MFCD00012930
CQNIYLLTIOPFCJ-UHFFFAOYSA-N
2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
2723909
(2,6-dichlorophenyl)hydrazine;hydrochloride
C1=CC(=C(C(=C1)Cl)NN)Cl.Cl
This item is not returnable. View return policy
50709-36-9
C6H7Cl3N2
213.486
MFCD00012930
CQNIYLLTIOPFCJ-UHFFFAOYSA-N
2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
2723909
(2,6-dichlorophenyl)hydrazine;hydrochloride
C1=CC(=C(C(=C1)Cl)NN)Cl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼225°C (decomposition)
Quantity 1 g
Beilstein 4569738
Formula Weight 213.5
Percent Purity ≥98%
Chemical Name or Material 2,6-Dichlorophenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'