missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenethyl alcohol, 96%, Thermo Scientific Chemicals

Catalog Number p-106370
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
30595-79-0
C8H8Cl2O
191.05
MFCD00800674
ZBQPKQUIKJDGIX-UHFFFAOYSA-N
2,6-dichlorophenethyl alcohol, 2-2,6-dichlorophenyl ethanol, 2,6-dichlorophenethylalcohol, 2-2,6-dichlorophenyl ethan-1-ol, 2,6-dichlorophenethanol, benzeneethanol, 2,6-dichloro, acmc-209hha, 2,6-dichlorobenzeneethanol, ksc493m0h, benzeneethanol,2,6-dichloro
2734100
2-(2,6-dichlorophenyl)ethanol
OCCC1=C(Cl)C=CC=C1Cl
This item is not returnable. View return policy
30595-79-0
C8H8Cl2O
191.05
MFCD00800674
ZBQPKQUIKJDGIX-UHFFFAOYSA-N
2,6-dichlorophenethyl alcohol, 2-2,6-dichlorophenyl ethanol, 2,6-dichlorophenethylalcohol, 2-2,6-dichlorophenyl ethan-1-ol, 2,6-dichlorophenethanol, benzeneethanol, 2,6-dichloro, acmc-209hha, 2,6-dichlorobenzeneethanol, ksc493m0h, benzeneethanol,2,6-dichloro
2734100
2-(2,6-dichlorophenyl)ethanol
OCCC1=C(Cl)C=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 59°C to 65°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 191.06
Percent Purity 96%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2, 6-Dichlorophenethyl alcohol, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'